1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

Lieferant: Apollo Scientific

PC200077-1G PC200077-500MG PC200077-5G
APOSPC200077-1GEA 270 EUR
APOSPC200077-1G APOSPC200077-500MG APOSPC200077-5G
1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile
1-(tert-Butyl)-6-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile
Formel: C₁₄H₁₄F₃N₃
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827797
CAS-Nummer: 1146221-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR