2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinamine

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2-(methylthio)-6-(trifluoromethyl)pyrimidine

PC200114-1G PC200114-500MG
APOSPC200114-1GEA 169 EUR
APOSPC200114-1G APOSPC200114-500MG
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinamine
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinamine
Formel: C₆H₆F₃N₃S
Molecular Weight: 209.19 g/mol
Schmelzpunkt: 120…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01011766
CAS-Nummer: 85730-32-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR