Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC200117-1G
PC200117-500MG
APOSPC200117-1GEA
757
EUR
APOSPC200117-1G
APOSPC200117-500MG
Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate
Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate
Formel:
C₉H₇ClF₃NO₂ Molecular Weight: 253,61 g/mol Siedepunkt: 74…76 °C/0,8mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD14581656 CAS-Nummer: 1198475-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...