Kalium(4-iodphenyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
(4-Iodphenyl)trifluorborsäure Kaliumsalz
PC200131-1G
PC200131-500MG
PC200131-5G
APOSPC200131-1GEA
118
EUR
APOSPC200131-1G
APOSPC200131-500MG
APOSPC200131-5G
Kalium(4-iodphenyl)trifluoroborat
Kalium(4-iodphenyl)trifluoroborat
Formel:
C₆H₄BF₃IK Molecular Weight: 309.91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800738 CAS-Nummer: 912350-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...