Kalium(3-iodphenyl)trifluoroborat

Lieferant: Apollo Scientific

Synonyme: Potassium 3-iodophenyltrifluoroborate

PC200132-1G PC200132-500MG PC200132-5G
APOSPC200132-1GEA 118 EUR
APOSPC200132-1G APOSPC200132-500MG APOSPC200132-5G
Kalium(3-iodphenyl)trifluoroborat
Potassium-3-iodophenyltrifluoroborate
Formel: C₆H₄BIF₃K
Molecular Weight: 309,91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800739
CAS-Nummer: 1189097-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR