Kalium(4-(phenylaminocarbonyl)phenyl)trifluoroborat

Lieferant: Apollo Scientific

PC200145-1G PC200145-5G
APOSPC200145-1GEA 87.3 EUR
APOSPC200145-1G APOSPC200145-5G
Kalium(4-(phenylaminocarbonyl)phenyl)trifluoroborat
Kalium(4-(phenylaminocarbonyl)phenyl)trifluoroborat
Formel: C₁₃H₁₀BF₃KNO
Molecular Weight: 303.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977719
CAS-Nummer: 912350-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR