7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

PC200146-1G PC200146-500MG
APOSPC200146-1GEA 210 EUR
APOSPC200146-1G APOSPC200146-500MG
7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one
7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one
Formel: C₈H₆FNO₂
Molecular Weight: 167,1372 g/mol
Schmelzpunkt: 203…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07774200
CAS-Nummer: 103361-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR