2-Chloro-5-fluoro-6-methoxynicotinic acid 90%

Lieferant: Apollo Scientific
Achtung

PC200151-1G PC200151-500MG
APOSPC200151-1GEA 270 EUR
APOSPC200151-1G APOSPC200151-500MG
2-Chloro-5-fluoro-6-methoxynicotinic acid 90%
2-Chloro-5-fluoro-6-methoxynicotinic acid
Formel: C₇H₅ClFNO₃
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12025843
CAS-Nummer: 943025-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR