8-Nitro-6-(trifluoromethyl)-4(1H)-quinolinone
Lieferant: Apollo Scientific
PC200172-1G
PC200172-500MG
APOSPC200172-1GEA
210
EUR
APOSPC200172-1G
APOSPC200172-500MG
8-Nitro-6-(trifluoromethyl)-4(1H)-quinolinone
8-Nitro-6-(trifluoromethyl)-4(1H)-quinolinone
Formel:
C₁₀H₅F₃N₂O₃ Molecular Weight: 258,16 g/mol Schmelzpunkt: 104…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12498705 CAS-Nummer: 190140-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...