6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile

Lieferant: Apollo Scientific

PC200181-1G PC200181-500MG
APOSPC200181-1GEA 311 EUR
APOSPC200181-1G APOSPC200181-500MG
6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Formel: C₁₀H₇F₃N₂O
Molecular Weight: 228,18 g/mol
Schmelzpunkt: 229…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04125115
CAS-Nummer: 861212-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR