2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone
Lieferant: Apollo Scientific
Synonyme:
3-(Trifluoroacetyl)-1H-pyrrole
PC200185-1G
PC200185-500MG
APOSPC200185-1GEA
386
EUR
APOSPC200185-1G
APOSPC200185-500MG
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone
Formel:
C₆H₄F₃NO Molecular Weight: 163.1 g/mol Schmelzpunkt: 110…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12924726 CAS-Nummer: 130408-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...