5-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
2-(5-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-Dioxaborolane, 2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-, 5-Fluoro-2-methoxyphenylboronic acid pinacol ester
PC200196-1G
PC200196-500MG
PC200196-5G
APOSPC200196-1GEA
118
EUR
APOSPC200196-1G
APOSPC200196-500MG
APOSPC200196-5G
5-Fluoro-2-methoxybenzeneboronic acid pinacol ester
5-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Formel:
C₁₃H₁₈BFO₃ Molecular Weight: 252.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16996296 CAS-Nummer: 1383806-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...