Methyl-3-(trifluoromethyl)pyridine-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC200197-1G
PC200197-500MG
APOSPC200197-1GEA
434
EUR
APOSPC200197-1G
APOSPC200197-500MG
Methyl-3-(trifluoromethyl)pyridine-4-carboxylate
Methyl-3-(trifluoromethyl)pyridine-4-carboxylate
Formel:
C₈H₆F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698095 CAS-Nummer: 1203952-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...