4-Fluoro-3-methylbenzeneboronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC200203-25G PC200203-5G
APOSPC200203-25GEA 271 EUR
APOSPC200203-25G APOSPC200203-5G
4-Fluoro-3-methylbenzeneboronic acid pinacol ester 98%
4-Fluoro-3-methylbenzeneboronic acid pinacol ester
Formel: C₁₃H₁₈BFO₂
Molecular Weight: 236,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18729904
CAS-Nummer: 1001200-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR