6-Bromo-3-(2,2,2-trifluoroethyl)-4(3H)-quinazolinone

Lieferant: Apollo Scientific

PC200214-1G PC200214-500MG
APOSPC200214-1GEA 671 EUR
APOSPC200214-1G APOSPC200214-500MG
6-Bromo-3-(2,2,2-trifluoroethyl)-4(3H)-quinazolinone
6-Bromo-3-(2,2,2-trifluoroethyl)-4(3H)-quinazolinone
Formel: C₁₀H₆BrF₃N₂O
Molecular Weight: 307,08 g/mol
Schmelzpunkt: 300…301 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD12025868
CAS-Nummer: 1135283-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR