3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine
Lieferant: Apollo Scientific
PC200215-1G
PC200215-500MG
PC200215-5G
APOSPC200215-1GEA
132
EUR
APOSPC200215-1G
APOSPC200215-500MG
APOSPC200215-5G
3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine
3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine
Formel:
C₁₂H₆Cl₃F₃N₂O Schmelzpunkt: 143…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11553026 CAS-Nummer: 73265-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...