3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-5-(trifluoromethyl)picolinonitrile
PC200219-1G
PC200219-5G
APOSPC200219-1GEA
27.7
EUR
APOSPC200219-1G
APOSPC200219-5G
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
Formel:
C₇H₂F₄N₂ Molecular Weight: 190,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848039 CAS-Nummer: 80194-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...