3-Ethynyl-5-(trifluoromethyl)-2-pyridinylamine

Lieferant: Apollo Scientific

PC200227-1G PC200227-500MG
APOSPC200227-1GEA 524 EUR
APOSPC200227-1G APOSPC200227-500MG
3-Ethynyl-5-(trifluoromethyl)-2-pyridinylamine
3-Ethynyl-5-(trifluoromethyl)-2-pyridinylamine
Formel: C₈H₅F₃N₂
Molecular Weight: 186.14 g/mol
Schmelzpunkt: 105…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100244
CAS-Nummer: 1048914-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR