Ethyl-1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylate

Lieferant: Apollo Scientific

PC200228-1G PC200228-500MG PC200228-5G
APOSPC200228-1GEA 132 EUR
APOSPC200228-1G APOSPC200228-500MG APOSPC200228-5G
Ethyl-1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylate
Ethyl-1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-3-carboxylate
Formel: C₁₄H₁₆ClF₃N₂O₂
Molecular Weight: 336.74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16890141
CAS-Nummer: 1242267-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR