1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile

Lieferant: Apollo Scientific

PC200229-1G PC200229-500MG PC200229-5G
APOSPC200229-1GEA 190 EUR
APOSPC200229-1G APOSPC200229-500MG APOSPC200229-5G
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile
Formel: C₁₂H₁₁ClF₃N₃
Molecular Weight: 289.69 g/mol
Schmelzpunkt: 108…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12498704
CAS-Nummer: 1171918-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR