[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3-(hydroxymethyl)-6-(trifluoromethyl)pyridine

PC200231-1G PC200231-500MG PC200231-5G
APOSPC200231-1GEA 328 EUR
APOSPC200231-1G APOSPC200231-500MG APOSPC200231-5G
[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol 95%
[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol
Formel: C₇H₅BrF₃NO
Molecular Weight: 256.03 g/mol
Schmelzpunkt: 67…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11848029
CAS-Nummer: 888738-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR