1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine
Lieferant: Apollo Scientific
PC200236-1G
PC200236-500MG
APOSPC200236-1GEA
270
EUR
APOSPC200236-1G
APOSPC200236-500MG
1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine
1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine
Formel:
C₁₁H₁₂F₃N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204721 CAS-Nummer: 175277-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...