5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine
Lieferant: Apollo Scientific
PC200240-1G
PC200240-500MG
APOSPC200240-1GEA
190
EUR
APOSPC200240-1G
APOSPC200240-500MG
5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine
5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine
Formel:
C₁₁H₈FN₃O₂ Schmelzpunkt: 255…258 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14581630 CAS-Nummer: 640271-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...