5-Nitro-6-(trifluoromethyl)-1H-indole

Lieferant: Apollo Scientific

PC200250-1G PC200250-500MG
APOSPC200250-1GEA 415 EUR
APOSPC200250-1G APOSPC200250-500MG
5-Nitro-6-(trifluoromethyl)-1H-indole
5-Nitro-6-(trifluoromethyl)-1H-indole
Formel: C₉H₅F₃N₂O₂
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827792
CAS-Nummer: 1186405-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR