6-Cyclopropyl-1-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

Lieferant: Apollo Scientific

Synonyme: 3-Amino-6-cyclopropyl-1-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine

PC200272-1G PC200272-500MG
APOSPC200272-1GEA 519 EUR
APOSPC200272-1G APOSPC200272-500MG
6-Cyclopropyl-1-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
6-Cyclopropyl-1-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
Formel: C₁₁H₁₁F₃N₄
Molecular Weight: 256.23 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16890143
CAS-Nummer: 1242267-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR