5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC200273-1G PC200273-500MG
APOSPC200273-1GEA 519 EUR
APOSPC200273-1G APOSPC200273-500MG
5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine
5-Methoxy-4-piperazino-2-(trifluoromethyl)pyrimidine
Formel: C₁₀H₁₃F₃N₄O
Molecular Weight: 262,24 g/mol
Schmelzpunkt: 101…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100216
CAS-Nummer: 1048912-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR