4-Methoxy-2-(methylsulphanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid

Lieferant: Apollo Scientific

PC200278-1G PC200278-500MG
APOSPC200278-1GEA 519 EUR
APOSPC200278-1G APOSPC200278-500MG
4-Methoxy-2-(methylsulphanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid
4-Methoxy-2-(methylsulphanyl)-6-(trifluoromethyl)-5-pyrimidinecarboxylic acid
Formel: C₈H₇F₃N₂O₃S
Molecular Weight: 268.22 g/mol
Schmelzpunkt: 158…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827810
CAS-Nummer: 1186404-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR