1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
Lieferant: Apollo Scientific
Synonyme:
1-(6-Chloro-5-(trifluoromethyl)pyridin-2-yl)piperazine
PC200280-1G
PC200280-500MG
APOSPC200280-1GEA
364
EUR
APOSPC200280-1G
APOSPC200280-500MG
1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine
Formel:
C₁₀H₁₁ClF₃N₃ Molecular Weight: 265.67 g/mol Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00892750 CAS-Nummer: 132834-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...