3-Bromo-4-fluoro-7-nitro-1H-indazole

Lieferant: Apollo Scientific

PC200290-1G PC200290-500MG
APOSPC200290-1GEA 626 EUR
APOSPC200290-1G APOSPC200290-500MG
3-Bromo-4-fluoro-7-nitro-1H-indazole
3-Bromo-4-fluoro-7-nitro-1H-indazole
Formel: C₇H₃BrFN₃O₂
Molecular Weight: 260.03 g/mol
Schmelzpunkt: 197…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20444429
CAS-Nummer: 1353877-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR