[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-(hydroxymethyl)-4-(trifluoromethyl)pyridine

PC200302-1G PC200302-500MG
APOSPC200302-1GEA 724 EUR
APOSPC200302-1G APOSPC200302-500MG
[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol
[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol
Formel: C₇H₅ClF₃NO
Molecular Weight: 211,57 g/mol
Siedepunkt: 140 °C/0,9mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD16622807
CAS-Nummer: 1228182-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR