3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid
Lieferant: Apollo Scientific
PC200303-1G
PC200303-500MG
APOSPC200303-1GEA
724
EUR
APOSPC200303-1G
APOSPC200303-500MG
3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid
3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid
Formel:
C₇H₃ClF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14581664 CAS-Nummer: 796090-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...