[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol

Lieferant: Apollo Scientific

Synonyme: 2,3-Dichloro-4-(hydroxymethyl)-5-(trifluoromethyl)pyridine

PC200304-1G PC200304-500MG
APOSPC200304-1GEA 724 EUR
APOSPC200304-1G APOSPC200304-500MG
[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol
[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol
Formel: C₇H₄Cl₂F₃NO
Molecular Weight: 246,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025911
CAS-Nummer: 1160474-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR