2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Bis(trifluoromethyl)isonicotinic acid

PC200305-1G PC200305-500MG
APOSPC200305-1GEA 1090 EUR
APOSPC200305-1G APOSPC200305-500MG
2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid
2,6-Bis(trifluoromethyl)pyridine-4-carboxylic acid
Formel: C₈H₃F₆NO₂
Molecular Weight: 259,11 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11501683
CAS-Nummer: 1092343-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR