2-Bromo-4-fluoro-5-hydroxybenzoic acid 90%

Lieferant: Apollo Scientific

PC200314-1G PC200314-500MG PC200314-5G
APOSPC200314-1GEA 270 EUR
APOSPC200314-1G APOSPC200314-500MG APOSPC200314-5G
2-Bromo-4-fluoro-5-hydroxybenzoic acid 90%
2-Bromo-4-fluoro-5-hydroxybenzoic acid
Formel: C₇H₄BrFO₃
Schmelzpunkt: 188…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD21333206
CAS-Nummer: 91659-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR