Methyl-4-fluoro-5-methoxy-2-nitrobenzoate

Lieferant: Apollo Scientific

Synonyme: Methyl 4-fluoro-5-methoxy-2-nitrobenzoate

PC200332-1G PC200332-500MG
APOSPC200332-1GEA 248 EUR
APOSPC200332-1G APOSPC200332-500MG
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate
Formel: C₉H₈FNO₅
Molecular Weight: 229.17 g/mol
Schmelzpunkt: 45…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20921594
CAS-Nummer: 159768-50-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR