N-[4-Iodo-2-(trifluoromethoxy)phenyl]acetamide

Lieferant: Apollo Scientific

PC200334-1G PC200334-500MG
APOSPC200334-1GEA 336 EUR
APOSPC200334-1G APOSPC200334-500MG
N-[4-Iodo-2-(trifluoromethoxy)phenyl]acetamide
N-[4-Iodo-2-(trifluoromethoxy)phenyl]acetamide
Formel: C₉H₇F₃INO₂
Molecular Weight: 345.06 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06408815
CAS-Nummer: 874814-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR