4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-3-fluorophenylboronic acid pinacol ester

PC200440-25G
APOSPC200440-25GEA 148 EUR
APOSPC200440-25G
4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%
4-Cyano-3-fluorobenzeneboronic acid pinacol ester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,076 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16996321
CAS-Nummer: 870238-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR