4-Fluoro-2-(1H-pyrazol-3-yl)phenol

Lieferant: Apollo Scientific

PC200541-1G PC200541-100MG PC200541-250MG PC200541-500MG PC200541-2.5G PC200541-5G
APOSPC200541-1GEA 180 EUR
APOSPC200541-1G APOSPC200541-100MG APOSPC200541-250MG APOSPC200541-500MG APOSPC200541-2.5G APOSPC200541-5G
4-Fluoro-2-(1H-pyrazol-3-yl)phenol
4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Formel: C₉H₇FN₂O
Molecular Weight: 178,17 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00104658
CAS-Nummer: 288401-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR