4-Chlor-6-fluor-2-methylchinolin

Lieferant: Apollo Scientific

PC200558-1G PC200558-5G
APOSPC200558-1GEA 52.2 EUR
APOSPC200558-1G APOSPC200558-5G
4-Chlor-6-fluor-2-methylchinolin
4-Chloro-6-fluoro-2-methylquinoline
Formel: C₁₀H₇ClFN
Molecular Weight: 195.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272332
CAS-Nummer: 18529-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR