4-Brom-2,6-difluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2006-100G
PC2006-25G
APOSPC2006-100GEA
55.1
EUR
APOSPC2006-100G
APOSPC2006-25G
4-Brom-2,6-difluoranilin 97%
4-Brom-2,6-difluoranilin
Formel:
C₆H₄BrF₂N Molecular Weight: 208,01 g/mol Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013389 CAS-Nummer: 67567-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...