2-Amino-5-fluorbenzothiazol
Lieferant: Apollo Scientific
Synonyme:
5-Fluorobenzo[d]thiazol-2-amine
PC200600-5G
APOSPC200600-5GEA
108
EUR
APOSPC200600-5G
2-Amino-5-fluorbenzothiazol
2-Amino-5-fluorobenzothiazole
Formel:
C₇H₅FN₂S Molecular Weight: 168,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664560 CAS-Nummer: 20358-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...