6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine 97%
Lieferant: Apollo Scientific
PC201013-1G
PC201013-250MG
APOSPC201013-1GEA
212
EUR
APOSPC201013-1G
APOSPC201013-250MG
6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine 97%
6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine
Formel:
C₈H₈FNO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
105655-00-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...