5-Methyl-2-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

PC201017-1G PC201017-5G
APOSPC201017-1GEA 82.8 EUR
APOSPC201017-1G APOSPC201017-5G
5-Methyl-2-(trifluoromethyl)aniline 95%
5-Methyl-2-(trifluoromethyl)aniline
Formel: C₈H₈F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD04972928
CAS-Nummer: 106877-29-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR