3-Brom-7-fluor-1H-indazol 97%

Lieferant: Apollo Scientific
Achtung

PC201097-1G PC201097-250MG PC201097-5G
APOSPC201097-1GEA 187 EUR
APOSPC201097-1G APOSPC201097-250MG APOSPC201097-5G
3-Brom-7-fluor-1H-indazol 97%
3-Bromo-7-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193285
CAS-Nummer: 1257853-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR