3-Brom-5-fluorchinolin 95+%

Lieferant: Apollo Scientific

PC201138-100MG PC201138-250MG
APOSPC201138-100MGEA 275 EUR
APOSPC201138-100MG APOSPC201138-250MG
3-Brom-5-fluorchinolin 95+%
3-Bromo-5-fluoroquinoline
Formel: C₉H₅BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD22690573
CAS-Nummer: 1416439-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR