2-Amino-4-chloro-5-fluoropyrimidine 97%
Lieferant: Apollo Scientific
PC201167-100MG
PC201167-1G
PC201167-250MG
APOSPC201167-100MGEA
85.8
EUR
APOSPC201167-100MG
APOSPC201167-1G
APOSPC201167-250MG
2-Amino-4-chloro-5-fluoropyrimidine 97%
2-Amino-4-chloro-5-fluoropyrimidine
Formel:
C₄H₃ClFN₃ Molecular Weight: 147,5400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09746440 CAS-Nummer: 1683-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...