3-Fluor-4-pyridon 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-4-hydroxypyridine, 3-Fluoropyridin-4-ol

PC201201-10G PC201201-1G PC201201-5G
APOSPC201201-10GEA 440 EUR
APOSPC201201-10G APOSPC201201-1G APOSPC201201-5G
3-Fluor-4-pyridon 95+%
3-Fluor-4-pyridon
Formel: C₅H₄FNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 22282-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR