7-Fluor-1H-indazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC201233-5G
PC201233-1G
APOSPC201233-5GEA
43.5
EUR
APOSPC201233-5G
APOSPC201233-1G
7-Fluor-1H-indazol 98%
7-Fluoro-1H-indazole
Formel:
C₇H₅FN₂ Molecular Weight: 136.1264 g/mol Schmelzpunkt: 123…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09026976 CAS-Nummer: 341-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...