4-Chloro-2-fluoropyridine 98%

Lieferant: Apollo Scientific

PC201236-100G PC201236-25G PC201236-5G
APOSPC201236-100GEA 439 EUR
APOSPC201236-100G APOSPC201236-25G APOSPC201236-5G
4-Chloro-2-fluoropyridine 98%
4-Chloro-2-fluoropyridine
Formel: C₅H₃ClFN
Molecular Weight: 131.53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112503
CAS-Nummer: 34941-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR