2-Chlor-3-fluor-6-methylpyridin 98%

Lieferant: Apollo Scientific

PC201240-10G PC201240-5G
APOSPC201240-10GEA 309 EUR
APOSPC201240-10G APOSPC201240-5G
2-Chlor-3-fluor-6-methylpyridin 98%
2-Chloro-3-fluoro-6-methylpyridine
Formel: C₆H₅ClFN
Molecular Weight: 145,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095216
CAS-Nummer: 374633-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR